Learn More
Mouse anti-Human IgG1, Clone: G17-1, biotin, Secondary Antibody, BD
Description
The G17-1 antibody reacts specifically with human IgG1. It does not react with other Ig isotypes. This antibody is routinely tested by ELISA analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G17-1
Isotype: IgG2b, κ
Species Reactivity: Human
ELISA
Specifications
Specifications
| Antigen | Mouse anti-Human IgG1 |
| Classification | Monoclonal |
| Clone | G17-1 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.