missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Human Ig, κ Light Chain, Secondary Antibody, BD

Product Code. 15812089
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 Tests
missing translation for 'unitSize':
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
15812089 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15812089 missing translation for 'mfr' BD Biosciences missing translation for 'supplierNo' 562619
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The G20-193 monoclonal antibody specifically binds to human immunoglobulin light chain, kappa (κ). It does not react with human immunoglobulin λ light chains or human immunoglobulin heavy chains.The antibody was conjugated to Brilliant Violet 421 and has been developed in collaboration with Sirigen. With an Ex Max of 407nm and Em Max at 421nm, Brilliant Violet 421 can be excited by the violet laser and detected in the standard Pacific Blue filter set (e.g., 450/50nm filter). Brilliant Violet 421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Host Species: Mouse
Clone: G20-193
Isotype: IgG1, κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Ig, κ Light Chain
Classification Monoclonal
Clone G20-193
Conjugate Brilliant Violet 421
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.