Learn More
Mouse anti-Human Ig κ Light Chain, Clone: G20-361, Secondary Antibody, BD
Description
The G20-361 antibody reacts with human immunoglobulin light chain kappa (κ). It does not react with λ light chain or heavy chains. This antibody is routinely tested by ELISA analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G20-361
Isotype: IgG2b, κ
Species Reactivity: Human
ELISA
Specifications
Specifications
| Antigen | Mouse anti-Human Ig κ Light Chain |
| Classification | Monoclonal |
| Clone | G20-361 |
| Concentration | 0.5mg/mL |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.