Learn More
CD45RA Mouse anti-Human, Brilliant Violet 650, Clone: 5H9, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 740608
Description
The 5H9 monoclonal antibody specifically binds to the human form of the leukocyte common antigen (LCA), CD45RA. This clone also crossreacts with CD45RA expressed by peripheral blood lymphocytes, monocytes and some granulocytes from rhesus macaque, baboon, and cynomolgus monkeys. The observed frequency of CD45RA positive lymphocytes is higher in the non-human primate compared to what is seen with normal human donor lymphocytes.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
| CD45RA | |
| Monoclonal | |
| 0.2mg/mL | |
| CD45R; PTPRC; LCA; Leukocyte common antigen | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 5H9 | |
| Brilliant Violet 650 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only