missing translation for 'onlineSavingsMsg'
Learn More

HSPD1, Mouse, Clone: LK2, Abnova™

Product Code. 16241630
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16241630 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16241630 Supplier Abnova Supplier No. MAB6622.200ug
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse monoclonal antibody raised against recombinant HSPD1.

This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Two pseudogenes, both located on chromosome 8, have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13. [provided by RefSeq

Specifications

Antigen HSPD1
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone LK2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant HSPD1.
Dilution Western Blot (0.25 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.1mM PMSF)
Gene HSPD1
Gene Alias CPN60/GROEL/HLD4/HSP60/HSP65/HuCHA60/SPG13
Gene Symbols HSPD1
Host Species Mouse
Immunogen Recombinant protein corresponding to human HSPD1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3329
Test Specificity Detects an ∼60KDa protein corresponding to the molecular mass of Hsp60 on SDS PAGE immunoblots.
Target Species Bovine, Canine, Chicken, E. coli, Guinea Pig, Hamster, H. pylori, Human, Monkey, Mouse, Nematode, Plant, Pig, Rabbit, Rat, S. typhi, Yeast
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.