missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR1, Mouse, PE, Clone: MEM-267, Abnova™
Mouse monoclonal antibody raised against HLA-DR1.
Brand: Abnova MAB5091.100ug
This item is not returnable.
View return policy
Specifications
| HLA-DR1 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Mouse monoclonal antibody raised against HLA-DR1. | |
| In PBS (0.2% BSA, 0.09% sodium azide) | |
| insoluble DR1 beta chain (DRB1*0101) expressed in Escherichia coli inclusion bodies., Purified | |
| RUO | |
| This antibody specifically binds to the empty but not peptide-loaded form of HLA-DR1. DR is the isotypes of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages). | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
| Flow Cytometry | |
| MEM-267 | |
| PE | |
| Flow Cytometry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction