missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Hamster IgG Cocktail, Clone: G94-90.5, G70-204, Secondary Antibody, BD

Product Code. 15817558
Change view
Click to view available options
Quantity:
0.2 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15817558 0.2 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15817558 Supplier BD Biosciences Supplier No. 554056
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

This preparation consists of a mixture of two mouse anti-hamster IgG monoclonal antibodies. It reacts with Armenian hamster IgG1/2/3 and Syrian hamster IgG1 monoclonal antibodies. It does not react with other hamster IgG groups and hamster IgM antibodies.

Host Species: Mouse
Clone: G94-90.5, G70-204
Isotype: IgG1, κ and IgG1, κ
Species Reactivity: Hamster
Immunogen: Pooled Armenian Hamster IgG mAb

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Hamster IgG Cocktail
Classification Monoclonal
Clone G70-204, G94-90.5
Conjugate PE
Host Species Mouse
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Hamster
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.