Learn More
Mouse, Anti-Glial Fibrillary Acidic Protein, Unconjugated, MP Biomedicals
Mouse Monoclonal Antibody
Brand: MP Biomedicals 08691101
Description
Product is a mouse monoclonal antibody, isotype IgG1, that reacts with Glial Fibrillary Acidic Protein.
Mouse Anti - Glial Fibrillary Acidic Protein is used in immunohistochemistry. It is designed for the specific and qualitative localization of GFAP in formalin-fixed paraffin-embedded tissue sections.
This antibody stains glial cells (Bergmann glia) and astrocytes. It has been tested for immunohistochemistry in human, pig, and rat. It is also suitable for microarray.
Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Key Applications: Immunoassays, Immunohistochemistry, Microarray
Specifications
| Immunoassay | |
| Unconjugated | |
| 50 μL | |
| Primary | |
| Liquid |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Multi-species |