Learn More
FGA, Mouse, Clone: 1F7, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of FGA.
Brand: Abnova MAB2948.200ug
Description
The protein encoded by this gene is the alpha component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia, afibrinogenemia and renal amyloidosis. Alternative splicing results in two isoforms which vary in the carboxy-terminus. [provided by RefSeq
Specifications
| FGA | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| FGA | |
| FGA | |
| A synthetic peptide corresponding to human FGA. | |
| RUO | |
| 2243 | |
| Human | |
| Liquid |
| ELISA | |
| 1F7 | |
| Mouse monoclonal antibody raised against synthetic peptide of FGA. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Fib2/MGC119422/MGC119423/MGC119425 | |
| Mouse | |
| 200 μg | |
| Primary | |
| This antibody reacts with Fibrinopeptide A region in the alpha-chain of fibrinogen and free fibrinopeptide A. | |
| Store at 4°C. | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.