Learn More
CD276 Mouse anti-Human, Brilliant Violet 421, Clone: 7-517, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565829
Description
The 7-517 monoclonal antibody specifically binds to CD276, also known as B7-H3 (B7 homolog 3). CD276 is a type I transmembrane glycoprotein and member of the B7 family of immunoregulatory proteins. Although the CD276 gene transcript is expressed in nearly all normal tissues, the protein is preferentially expressed in tumors, tumor cell lines, sinonasal epithelial cells, and extravillous trophoblast and Hofbauer cells of the placenta. CD276 protein expression can be induced by activation of T and B lymphocytes, natural killer (NK) cells and antigen-presenting cells. The roles of CD276 in the regulation of immune responses, especially to cancer, are under investigation.
Specifications
| CD276 | |
| Monoclonal | |
| 0.2mg/mL | |
| B7-H3, B7H3, 4Ig-B7-H3, B7RP-2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence | |
| 7-517 | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Monocyte-Derived Dendritic Cells | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.