missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Bovine IgG (H+L) Secondary Antibody (IL-A2), FITC, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Monoclonal antibody specifically detects IgG (H+L) in Bovine samples. It is validated for ELISA, Immunoprecipitation
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | IL-A2 |
| Conjugate | FITC |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Purified bovine IgG |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?