missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-ASGPR1, Clone: 8D7, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
739.00€
Specifications
| Antigen | ASGPR1 |
|---|---|
| Clone | 8D7 |
| Concentration | 0.1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay |
| Applications | Western Blot |
Description
ASGPR1 Monoclonal specifically detects ASGPR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| ASGPR1 | |
| 0.1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 0.2 um filtered solution in PBS and 0.1% BSA with 0.02% Sodium Azide | |
| 432 | |
| Crude rat liver membrane extracts | |
| Primary | |
| Store at 4°C. Do not freeze. |
| 8D7 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay | |
| Monoclonal | |
| Mouse | |
| Human | |
| ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 | |
| ASGR1 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title