missing translation for 'onlineSavingsMsg'
Få mere at vide

CD11c Mouse anti-Human, Unlabeled, Clone: 3.9, BD

Artikelnummer. 15844539
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.1 mg
Pakningsstørrelse:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
15844539 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15844539 Leverandør BD Biosciences Leverandørnr. 565805
 Restricted Item
Estimated Shipment: 30-07-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

The 3.9 monoclonal antibody specifically binds to CD11c, which is also known as Integrin alpha X ( α X Integrin/ITGAX), or p150,95 Integrin alpha chain. CD11c is a ∽150 kDa type I transmembrane glycoprotein. It is expressed on monocytes, macrophages, granulocytes, NK cells, dendritic cells, and subsets of B and T cells. It associates with CD18 (Integrin beta 2/ β 2 Integrin) to form the CD11c/CD18 complex, which is also known as p150,95 Integrin, or the Type 4 Complement Receptor (CR4). CD11c/CD18 binds fibrinogen and reportedly serves as a receptor for iC3b and ICAM-1/CD54. CD11c/CD18 functions as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium. The 3.9 monoclonal antibody crossreacts with Rhesus macaque leucocytes.

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen CD11c
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 3.9
Concentration 0.5mg/mL
Conjugate Unconjugated
Description ITGAX; AlphaX Integrin; Axb2; Integrin alpha-X; CR4; SLEB6; p150,95 alpha
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human monocytes and synovial cells
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.