missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOS Antibody (S3.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2784-0.025mg
This item is not returnable.
View return policy
Description
MOS Monoclonal specifically detects MOS in Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| MOS | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| MOS | |
| Recombinant (Xenopus laevis) Mos fusion protein | |
| 0.025 mg | |
| Primary | |
| Able to immunoprecipitate MOS from cell/egg extracts. Cross-reacts with Xenopus laevis. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot | |
| S3.1 | |
| Unconjugated | |
| c-mos, EC 2.7.11, EC 2.7.11.1, MGC119962, MGC119963, MSV, oncogene MOS, Moloney murine sarcoma virus, Oocyte maturation factor mos, Proto-oncogene c-Mos, proto-oncogene serine/threonine-protein kinase mos, v-mos Moloney murine sarcoma viral oncogene homolog | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 4342 | |
| Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction