missing translation for 'onlineSavingsMsg'
Learn More

MOS Antibody (S3.1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB100-2784AF700

Product Code. 18006034

  • 437.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MOS Monoclonal specifically detects MOS in Xenopus samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MOS
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
MOS
Recombinant (Xenopus laevis) Mos fusion protein
0.1 mL
Cell Cycle and Replication
4342
Store at 4C in the dark.
IgG2a
Western Blot, Immunohistochemistry
S3.1
Western Blot, Immunohistochemistry
c-mos, EC 2.7.11, EC 2.7.11.1, MGC119962, MGC119963, MSV, oncogene MOS, Moloney murine sarcoma virus, Oocyte maturation factor mos, Proto-oncogene c-Mos, proto-oncogene serine/threonine-protein kinase mos, v-mos Moloney murine sarcoma viral oncogene homolog
Mouse
Protein A purified
RUO
Primary
Xenopus species
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.