missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ MOPSO, 0.2M buffer soln., pH 7.0
37.94€ - 56.00€
Chemical Identifiers
| CAS | 79803-73-9 |
|---|---|
| Molecular Formula | C7H14NNaO5S |
| Molecular Weight (g/mol) | 247.241 |
| MDL Number | MFCD00070007 |
| InChI Key | WSFQLUVWDKCYSW-UHFFFAOYSA-M |
| PubChem CID | 23675131 |
| IUPAC Name | sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate |
| SMILES | C1COCCN1CC(CS(=O)(=O)[O-])O.[Na+] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
| 15414069 SDS |
Alfa Aesar™
J61469.AE |
100mL |
37.94€ 100mL |
from Supplier 09-05-2019 |
15424069 SDS |
Alfa Aesar™
J61469.AK |
250mL |
56.00€ 250mL |
from Supplier 09-05-2019 |
Chemical Identifiers
| 79803-73-9 | |
| 247.241 | |
| WSFQLUVWDKCYSW-UHFFFAOYSA-M | |
| sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate |
| C7H14NNaO5S | |
| MFCD00070007 | |
| 23675131 | |
| C1COCCN1CC(CS(=O)(=O)[O-])O.[Na+] |
Specifications
| MOPSO buffer soln. | |
| C7H14NNaO5S | |
| 0.2M buffer soln. | |
| C1COCCN1CC(CS(=O)(=O)[O-])O.[Na+] | |
| 247.241 | |
| Liquid | |
| 100mL |
| 79803-73-9 | |
| MFCD00070007 | |
| WSFQLUVWDKCYSW-UHFFFAOYSA-M | |
| sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate | |
| 23675131 | |
| 7.0 |