Learn More
Montelukast Ethyl Ester, Mikromol™
Description
Montelukast Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Montelukast Ethyl Ester |
| CAS | 1044705-96-5 |
| Analyte or Component Names | Montelukast Ethyl Ester |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C37 H40 Cl N O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Montelukast Ethyl Ester, 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-cyclopropaneacetic acid ethyl ester |
| InChI Formula | InChI=1S/C37H40ClNO3S/c1-4-42-35(40)24-37(20-21-37)25-43-34(19-15-27-9-5-6-11-32(27)36(2,3)41)29-10-7-8-26(22-29)12-17-31-18-14-28-13-16-30(38)23-33(28)39-31/h5-14,16-18,22-23,34,41H,4,15,19-21,24-25H2,1-3H3/b17-12+/t34-/m1/s1 |
| SMILES | CCOC(=O)CC1(CS[C@H](CCc2ccccc2C(C)(C)O)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3)CC1 |
| IUPAC Name | ethyl 2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetate |
| Show More |