missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Monoglyceride Lipase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30968-100ug
This item is not returnable.
View return policy
Description
Monoglyceride Lipase Polyclonal antibody specifically detects Monoglyceride Lipase in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Monoglyceride Lipase | |
| Polyclonal | |
| Western Blot : 1-2 μg/mL, Immunohistochemistry : 2-4 μg/mL | |
| HU-K5EC 3.1.1.23, Lysophospholipase homolog, Lysophospholipase-like, MAGL, MGLHUK5, Monoacylglycerol lipase, monoglyceride lipase | |
| Synthetic peptide derived from the human Monoglyceride Lipase protein. | |
| 100 μg | |
| Lipid and Metabolism | |
| 11343 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction