missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoamine Oxidase B Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
166.00€
Specifications
| Antigen | Monoamine Oxidase B |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.1 - 0.3 μg/mL, Flow Cytometry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Peptide ELISA Detection limit 1:32000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Monoamine Oxidase B Polyclonal specifically detects Monoamine Oxidase B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
| Monoamine Oxidase B | |
| Western Blot 0.1 - 0.3 μg/mL, Flow Cytometry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Peptide ELISA Detection limit 1:32000 | |
| Polyclonal | |
| Goat | |
| Human | |
| NP_000889.3 | |
| 4129 | |
| Peptide with sequence C-HKARKLARLTKEE corresponding to internal region according to NP_000889.3. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| adrenalin oxidase, amine oxidase [flavin-containing] B, EC 1.4.3, EC 1.4.3.4, MAO, brain, MAO, platelet, MAO-B, MGC26382, monoamine oxidase B, Monoamine oxidase type B, tyramine oxidase | |
| MAOB | |
| IgG | |
| Affinity Purified | |
| This antibody is not expected to cross-react with MAOA. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title