Learn More
Mono element - Hg for ICP/MS 1mg/L, Fisher chemical™
Product Code. 12986203 Shop All Fisher Chemical Products
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
50mL
Product Code. 12986203
Brand: Fisher Chemical MS 013 050
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Chemical Identifiers
CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
Specifications
Packaging Bottle
Physical Form Liquid
Concentration 1 mg/L
Type ICP/MS Calibration Multi-element Standard Solution
Solvent or Matrix 2% HNO3
Quantity 50 mL
Chemical Name or Material Mercury