missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Monkeypox Virus M1R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
218.00€ - 419.00€
Specifications
| Antigen | Monkeypox Virus M1R |
|---|---|
| Dilution | Western Blot 1:5000-1:20000, ELISA 1:5000-1:10000 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30042678
|
Novus Biologicals™
NBP3-30756-100ul |
100 μL |
419.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043011
|
Novus Biologicals™
NBP3-30756-20ul |
20 μL |
218.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Monkeypox Virus M1R Polyclonal antibody specifically detects Monkeypox Virus M1R in Virus samples. It is validated for ELISA,Western BlotSpecifications
| Monkeypox Virus M1R | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Produced in rabbits immunized with purified, recombinant Monkeypox Virus M1R (RefSeq#: QNP13675.1; Ala3-Gly183) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot 1:5000-1:20000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title