missing translation for 'onlineSavingsMsg'
Learn More

Monkeypox Virus I1L Antibody [mFluor Violet 500 SE], Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-43764MFV500

Product Code. 30281365

  • 529.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Monkeypox Virus I1L Polyclonal antibody specifically detects Monkeypox Virus I1L in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Monkeypox Virus I1L
Polyclonal
Western Blot
Rabbit
Antigen and protein A Affinity-purified
RUO
Virus
Purified
Western Blot
mFluor Violet 500 SE
50 mM Sodium Borate
Produced in rabbits immunized with purified, recombinant Monkeypox Virus I1L (Met1-Glu312)
0.1 mL
Primary
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.