missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus B21R Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
173.00€ - 451.00€
Specifications
| Antigen | Monkeypox Virus B21R |
|---|---|
| Dilution | Western Blot 1:2000-1:20000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30035140
|
Novus Biologicals
NBP3-26104-20ul |
20 μL |
173.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30036857
|
Novus Biologicals
NBP3-26104-100ul |
100 μL |
451.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Monkeypox Virus B21R Polyclonal antibody specifically detects Monkeypox Virus B21R in Virus samples. It is validated for Western BlotSpecifications
| Monkeypox Virus B21R | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Monkeypox Virus B21R | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:2000-1:20000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.0 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title