missing translation for 'onlineSavingsMsg'
Learn More

Molindone.HCl, 1mg/ml in Methanol (as free base), Lipomed

Product Code. 30464269
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30464269

Brand: Lipomed LPMMOL1886HC1LM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Molindone.HCl, 1mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Molindone Hydrochloride
CAS 15622-65-8
Analyte or Component Names Molindone Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H24 N2 O2 . Cl H
Concentration 1 mg/mL
Synonym 4 H-Indol-4-one, 3-ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-, hydrochloride (1:1), 4 H-Indol-4-one, 3-ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-, monohydrochloride (9 CI), Indol-4(5 H)-one, 3-ethyl-6,7-dihydro-2-methyl-5-(morpholinomethyl)-, monohydrochloride (8 CI), (±)-Molindone hydrochloride, EN 1733 A, Lindone, Moban, Molindone hydrochloride
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CCc1c(C)[nH]c2CCC(CN3CCOCC3)C(=O)c12
IUPAC Name 3-ethyl-2-methyl-5-(morpholin-4-ylmethyl)-1,5,6,7-tetrahydroindol-4-one;hydrochloride
Solvent or Matrix Methanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 312.83
Quantity 1 mL
Formula Weight 312.1605 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.