Learn More
Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Product Code. 12563647 Shop All Epredia Products
Click to view available options
Type:
Fast Green Stain Solution
Gold Chloride Solution (0.1%)
Methenamine/Borax
Periodic Acid Schiff
Silver Nitrate Solution (5%)
Silver Stain Kit, Periodic Acid
Sodium Thiosulfate Solution
Unit Size:
125mL
30mL
Each
Pack of 12
Product Code. 12563647
Brand: Epredia™ 88024
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
  • Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
  • Background is stained green
TRUSTED_SUSTAINABILITY
Specifications
Type Fast Green Stain Solution
Quantity 125 mL