missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mNeonGreen Mouse, Unconjugated, Clone: 32F6, Proteintech ChromoTek
Description
Mouse monoclonal antibody [32F6] to mNeonGreen fluorescent protein derived from Branchiostoma lanceolatum.
Specifications
Specifications
| Antigen | mNeonGreen |
| Applications | Western Blot, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Clone | 32F6 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Content And Storage | Shipped at ambient temperature. Store at +4°C. Stable for one year. |
| Isotype | IgG2c |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?