missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNDA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13576-1-AP-150UL
This item is not returnable.
View return policy
Description
Until recently, immunological markers for myeloid cells have been lacking, especially those which identify different levels of cellular differentiation. This antibody stains early precursor and mature forms of human myeloid cells. This mAB reacts with ant
Until recently, immunological markers for myeloid cells have been lacking, especially those which identify different levels of cellular differentiation. This antibody stains early precursor and mature forms of human myeloid cells. This mAB reacts with antigenic determits present in normal myeloid cells and leukemias of similar derivation.Specifications
| MNDA | |
| Polyclonal | |
| Unconjugated | |
| MNDA | |
| MNDA, PYHIN3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4332 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P41218 | |
| MNDA | |
| MNDA Fusion Protein Ag4476 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction