missing translation for 'onlineSavingsMsg'
Learn More

MNDA Mouse anti-Human, Alexa Fluor™ 647, Clone: 253 (also known as 253A), BD Pharmingen™

Product Code. 15805169
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15805169 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15805169 Supplier BD Biosciences Supplier No. 566582
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MNDA Monoclonal antibody specifically detects MNDA in Human samples. It is validated for Flow Cytometry,Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MNDA
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 253 (also known as 253A)
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.