Learn More
MMP19 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-14244-100UL
 View more versions of this product
Product Code. 18079475
  • 450.00€ / 100µL
Description
The MMP19 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MMP19. This antibody recognizes human antigen. The MMP19 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
MMP19
1000 μg/mL
matrix metallopeptidase 19, Matrix metalloproteinase 18, Matrix metalloproteinase 19, Matrix metalloproteinase RASI, MMP 18, MMP 19, MMP18, MMP19, RASI, RASI 1
100 μL
4327
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa