Learn More
MMP19 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-14244-20UL
Description
The MMP19 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MMP19. This antibody recognizes human antigen. The MMP19 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| MMP19 | |
| 1000 μg/mL | |
| matrix metallopeptidase 19, Matrix metalloproteinase 18, Matrix metalloproteinase 19, Matrix metalloproteinase RASI, MMP 18, MMP 19, MMP18, MMP19, RASI, RASI 1 | |
| 20 μL | |
| 4327 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |