Learn More
MMP-9 Antibody (CL0542) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52950AF532
Product Code. 30288091
  • 737.00€ / 0.10mL
Description
MMP-9 Monoclonal antibody specifically detects MMP-9 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MMP-9
Monoclonal
Alexa Fluor 532
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
This antibody was developed using a recombinant protein derived from P14780, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR
4318
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL0542
50mM Sodium Borate