Learn More
Novus Biologicals™ MMP-9 Antibody (andecaliximab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28078-50ug
 View more versions of this product
Product Code. 30045057
  • 213.00€ / 50µg
Description
MMP-9 Monoclonal antibody specifically detects MMP-9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MMP-9
Monoclonal
Unconjugated
P14780
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG4
Flow Cytometry, ELISA, Functional Assay
andecaliximab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
MMP9
50 μg
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR
4318