missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MMP-11 Antibody (SR3093), Novus Biologicals™
Rabbit Monoclonal Antibody
316.00€ - 382.00€
Specifications
| Antigen | MMP-11 |
|---|---|
| Clone | SR3093 |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041728
|
Novus Biologicals™
NBP3-27748-100ul |
100 μL |
382.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041781
|
Novus Biologicals™
NBP3-27748-50ul |
50 μL |
316.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-11 Monoclonal antibody specifically detects MMP-11 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| MMP-11 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.24, matrix metallopeptidase 11 (stromelysin 3), matrix metalloproteinase 11 (stromelysin 3), Matrix metalloproteinase-11, MMP-11, SL-3, ST3, STMY3stromelysin 3, stromelysin III, stromelysin-3 | |
| A synthesized peptide derived from human MMP-11 (Uniprot #: P24347) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3093 | |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 4320 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title