missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MLL3 Antibody (6D1B9), Novus Biologicals™
Mouse Monoclonal Antibody
287.00€ - 511.00€
Specifications
| Antigen | MLL3 |
|---|---|
| Clone | 6D1B9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30031245
|
Novus Biologicals™
NBP3-27147-0.1ml |
0.1 mL |
511.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30036418
|
Novus Biologicals™
NBP3-27147-0.025ml |
0.025 mL |
287.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLL3 Monoclonal antibody specifically detects MLL3 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ ImmunofluorescenceSpecifications
| MLL3 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| ALR-like protein, DKFZp686C08112, EC 2.1.1.43, FLJ12625, HALRMGC119851, histone-lysine N-methyltransferase MLL3, histone-lysine N-methyltransferase, H3 lysine-4 specific, Homologous to ALR protein, KIAA1506FLJ38309, KMT2C, KMT2CMGC119852, Lysine N-methyltransferase 2C, MGC119853, myeloid/lymphoid or mixed-lineage leukemia 3, Myeloid/lymphoid or mixed-lineage leukemia protein 3 | |
| Purified recombinant fragment of human MLL3 (AA: 1-205) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 6D1B9 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 58508 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title