missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLH3 Antibody (CL14096) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24597C
This item is not returnable.
View return policy
Description
MLH3 Monoclonal antibody specifically detects MLH3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| MLH3 | |
| Monoclonal | |
| DyLight 650 | |
| DNA mismatch repair protein Mlh3, HNPCC7, MGC138372, mutL (E. coli) homolog 3, mutL homolog 3 (E. coli), MutL protein homolog 3 | |
| This antibody was developed using a synthetic peptide derived from Q9UHC1, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| DNA Repair, Mismatch Repair | |
| 27030 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL14096 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction