missing translation for 'onlineSavingsMsg'
Learn More

MKX Monoclonal Antibody (OTI6A12), TrueMAB™, OriGene

Product Code. 17529763
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17529763 100 μL 100µL
17549763 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17529763 Supplier Origene Technologies Supplier No. TA808466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MKX is a member of an Iroquois (IRX) family-related class of ′three-amino acid loop extension′ (TALE) atypical homeobox proteins characterized by 3 additional amino acids in the loop region between helix I and helix II of the homeodomain.

Specifications

Antigen MKX
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6A12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MKX
Gene Accession No. Q8IYA7
Gene Alias C10orf48, IFRX, IRXL1
Gene Symbols MKX
Host Species Mouse
Immunogen Full length human recombit protein of human MKX produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283078
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.