missing translation for 'onlineSavingsMsg'
Learn More

MKX Monoclonal Antibody (OTI4H10), TrueMAB™, OriGene

Product Code. p-200018503
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17559513 30 μL 30µL
17539513 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17559513 Supplier Origene Technologies Supplier No. TA808350S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MKX Monoclonal antibody specifically detects MKX in Human samples. It is validated for Western Blot

Specifications

Antigen MKX
Applications Western Blot
Classification Monoclonal
Clone OTI4H10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Tp53rk
Gene Accession No. Q96S44
Gene Alias 9430023B20Rik; C10orf48; homeobox protein mohawk; IFRX; Iroquois family related homeodomain protein; iroquois homeobox protein-like 1; Irxl1; MKX; mohawk homeobox; RGD1305652
Host Species Mouse
Immunogen Full length human recombinant protein of human TP53RK produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112858
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.