missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKL2 Antibody (CL1546) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52970JF669
This item is not returnable.
View return policy
Description
MKL2 Monoclonal antibody specifically detects MKL2 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| MKL2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| DKFZp686J1745, FLJ31823, FLJ45623, KIAA1243, megakaryoblastic leukemia 2, MKL/myocardin-like 2, MKL/myocardin-like protein 2, MRTFB, MRTF-B, myocardin-related transcription factor B, NPD001 | |
| This antibody was developed using a recombinant protein derived from Q9ULH7, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cardiovascular Biology, Stem Cells | |
| 57496 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL1546 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction