missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MIOS Monoclonal Antibody (955413)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA562013

Product Code. 30535487

  • 492.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

As a component of the GATOR subcomplex GATOR2, functions within the amino acid-sensing branch of the TORC1 signaling pathway. Indirectly activates mTORC1 and the TORC1 signaling pathway through the inhibition of the GATOR1 subcomplex (PubMed:23723238). It is negatively regulated by the upstream amino acid sensors SESN2 and CASTOR1 (PubMed:25457612, PubMed:27487210). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MIOS
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q8VE19, Q9NXC5
MIOS
Human MIOS synthetic peptide.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1
Western Blot
955413
Unconjugated
MIOS
C81488; FLJ20323; GATOR complex protein MIOS; H_DJ1159O04.1; meiosis regulator for oocyte development; MIO; Mios; Missing oocyte meiosis regulator homolog; missing oocyte, meiosis regulator, homolog; missing oocyte, meiosis regulator, homolog (Drosophila); RGD1308432; Sea4; WD repeat-containing protein mio; WUGSC:H_DJ1159O04.1
Mouse
Protein A/G
RUO
252875, 362324, 54468
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.