Learn More
BOLA Miniature Screw-in Adaptors
Click to view available options
Connections:
Female
Male
Diameter (Metric) Inner:
0.8 mm
1.6 mm
Unit Size:
Each
Description
Adaptor made of PTFE for transition from female thread UNF ¼ in. 28 G to male thread NPT 1/8 in. or NPT ¼ in..
Specifications
Specifications
| Diameter (Metric) Outer | 1.6 mm |
| Connections | Female |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 0.8 mm |