Learn More
BOLA Miniature Screw-in Adaptors
Product Code. 10459191
Click to view available options
Connections:
Female
Male
Diameter (Metric) Inner:
0.8 mm
1.6 mm
Unit Size:
Each
Product Code. 10459191
Brand: BOLA F71602
Only null left
Add to basket
Add to basket
Miniature Screw-in Adaptors PTFE, UNF 1/4 28G female, NPT 1/4 in.
Adaptor made of PTFE for transition from female thread UNF ¼ in. 28 G to male thread NPT 1/8 in. or NPT ¼ in..
Specifications
Diameter (Metric) Outer 1.6 mm
Connections Female
Autoclavable Autoclavable
Diameter (Metric) Inner 0.8 mm