Learn More
BOLA Miniature Couplings
Product Code. 10378751
Click to view available options
Connections:
3
4
Diameter (Metric) Inner:
0.8 mm
1.6 mm
Unit Size:
Each
Product Code. 10378751
Brand: BOLA F70718
Only null left
Add to basket
Add to basket
Miniature Couplings PTFE
  • Three types of couplings available: straight type made of PTFE or POM with higher mechanical strength
  • T type with three connections or cross type with four connections, all made of PTFE.
  • The tubing is connected by means of tube end fittings to a female thread UNF ¼ in. 28 G.
Specifications
Diameter (Metric) Outer 1.6 mm
Connections 4
Autoclavable Autoclavable
Diameter (Metric) Inner 0.8 mm