missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mind Bomb 1/MIB1 Antibody (6A9C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61864
This item is not returnable.
View return policy
Description
Mind Bomb 1/MIB1 Monoclonal specifically detects Mind Bomb 1/MIB1 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Mind Bomb 1/MIB1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| DAPK-interacting protein 1, DIP1, DIP-1FLJ90676, DKFZp686I0769, DKFZp761M1710, E3 ubiquitin-protein ligase MIB1, EC 6.3.2, EC 6.3.2.-, KIAA1323, MGC129660, MIB, Mind bomb homolog 1, mindbomb homolog 1 (Drosophila), ubiquitin ligase mind bomb, ubiquitin ligase protein MIB1, Zinc finger ZZ type with ankyrin repeat domain protein 2, ZZANK2MGC129659, ZZZ6 | |
| Mouse | |
| 110 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Monkey | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 6A9C9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| MIB1 | |
| Purified recombinant fragment of human MIB1 (AA: 6-221) expressed in E. Coli. | |
| Protein G purified | |
| Lipid and Metabolism, Zinc Finger | |
| 57534 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction