Learn More
Milrinone, Mikromol™
Description
Milrinone
Specifications
Specifications
| Chemical Name or Material | Milrinone |
| CAS | 78415-72-2 |
| Analyte or Component Names | Milrinone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H9 N3 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Milrinone, [3,4'-Bipyridine]-5-carbonitrile, 1,6-dihydro-2-methyl-6-oxo-, 1,6-Dihydro-2-methyl-6-oxo-(3,4'-bipyridine)-5-carbonitrile, Corotrop, Corotrope, Milrinone, Primacor IV, Win 47203 |
| InChI Formula | InChI=1S/C12H9N3O/c1-8-11(9-2-4-14-5-3-9)6-10(7-13)12(16)15-8/h2-6H,1H3,(H,15,16) |
| SMILES | CC1=C(C=C(C#N)C(=O)N1)c2ccncc2 |
| IUPAC Name | 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile |
| Show More |