missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Midkine Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11009-1-AP-20UL
This item is not returnable.
View return policy
Description
Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.Specifications
| Midkine | |
| Polyclonal | |
| Unconjugated | |
| MDK | |
| ARAP, MDK, Midkine, MK, MK1, NEGF2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4192 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P21741 | |
| MDK | |
| Midkine Fusion Protein Ag1465 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction