Learn More
Poulten Graf™ Micropipettor Tip
Product Code. p-8006243
Click to view available options
Color:
Blue
Yellow
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 1000
Pack of 400
Product Code. 10381262
Brand: Poulten Graf™ D590
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Provides volumetric accuracy for your pipetting needs. Poulten Graf™ Micropipette Tips are calibrated and are available in standard sizes.
  • Exceptional moulded quality
  • Excellent fit for accurate working
  • Suitable for most leading brands
Specifications
Volume (Metric) 1 to 200 μL
Sterility Sterile
Color Yellow
Accuracy >0.75%
Certifications/Compliance Calibrated