Learn More
Socorex™ Micropipette Rack for Qualitix™ Tipfill Refill System
Product Code. 15790649
Change view
Click to view available options
Holds:
Micropipette Tips (10μL), Micropipette Tips (200μL )
Micropipette Tips (1000μL)
No. per Pack:
6
10
Unit Size:
Pack of 10
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds No. per Pack Unit Size
15790649 Micropipette Tips (1000μL) 6 Pack of 6
15770649 Micropipette Tips (10μL), Micropipette Tips (200μL ) 10 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15790649 Supplier Socorex™ Supplier No. 3070.1000ER
  • ONLINE EXCLUSIVE PRICE
    31.40€
    26.23€ / Pack of 6
    Save 5.17€
    16% Off
View all promotions
Estimated Shipment: 24-12-2026
Reusable, reclosable, and more space-saving than conventional racks. Socorex™ Micropipette Rack for Qualitix™ Tipfill Refill System readily accepts micropipette refill packs and has smooth, rounded edges and an ergonomic snap latch that won't catch gloves.
  • Accepts Qualitix™ Tipfill Refill System refill packs.
  • Hinged lid.
  • Smooth, rounded edges.
  • Ergonomic snap latch that won't catch gloves.
  • Transparent lid.
  • Reusable.
Specifications
Closure Included Yes
Autoclavable Autoclavable
Color Natural
Holds Micropipette Tips (1000μL)
Stackable Yes
For Use With (Equipment) Qualitix™Tipfill Refill System
No. per Case 8
No. per Pack 6
Array 96
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.