Learn More
Microcystin-LR Antibody (MC10E7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-89027F
This item is not returnable.
View return policy
Description
Microcystin-LR Monoclonal antibody specifically detects Microcystin-LR in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Microcystin-LR | |
| Monoclonal | |
| FITC | |
| PBS | |
| Microcystin-LR linked via N-methyl-dehydroalanine to cationized ovalbumin. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MC10E7 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Non-species specific | |
| Purified |