Learn More
Michigan Residual Solvents Mixture 471 1000 μg/mL in Triacetin, Dr. Ehrenstorfer
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
Specifications
Specifications
| CAS | 75-28-5, 106-97-8, 463-82-1, 74-98-6 |
| Quantity | 5 x 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Triacetin |