Learn More
Cyhalothrin 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30326378
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 30326378
Brand: Dr. Ehrenstorfer DREA11859450AL1000
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cyhalothrin
CAS 68085-85-8
Analyte or Component Names Cyhalothrin
Shipping Condition +20°C
Molecular Formula C23 H19 Cl F3 N O3
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propen-1-yl)-2,2-dimethyl-, cyano(3-phenoxyphenyl)methyl ester, Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, cyano(3-phenoxyphenyl)methyl ester (9CI), Clocythrin, Coopertix, Cyhalothrin, Gongfu, Grenade, ICI-PP 563, PP 563, Saber, α-Cyano-3-phenoxybenzyl 3-(2-chloro-3,3,3-trifluoroprop-1-en-1-yl)-2,2-dimethylcyclopropanecarboxylate, α-Cyhalothrin
InChI Formula InChI=1S/C23H19ClF3NO3/c1-22(2)17(12-19(24)23(25,26)27)20(22)21(29)31-18(13-28)14-7-6-10-16(11-14)30-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3
SMILES CC1(C)C(C=C(Cl)C(F)(F)F)C1C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2
IUPAC Name [cyano-(3-phenoxyphenyl)methyl] 3-(2-chloro-3,3,3-trifluoroprop-1-enyl)-2,2-dimethylcyclopropane-1-carboxylate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 449.85
Quantity 1 mL
Formula Weight 449.1006 g/mol